Learn More
Medchemexpress LLC HY-12212 100mg , RTA-408 Omaveloxolone CAS:1474034-05-3 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY12212100MG
Encompass_Preferred
Medchemexpress, HY-12212 100mg RTA-408 Omaveloxolone CAS:1474034-05-3 Formula:C33H44F2N2O3 Nrf2 Purity:98% RTA-408 is an antioxidant inflammation modulator (AIM), which activates Nrf2 and suppresses nitric oxide (NO). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3275
 Restricted Item