Learn More
TARGETMOL CHEMICALS INC RTC-5 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1277710MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RTC-5 (TRC-382) (TRC-382) is a phenothiazine compound that has been specifically enhanced for its potent anti-cancer properties. It exhibits considerable effectiveness against an EGFR-driven cancer xenograft model. The mechanism behind its efficacy can be attributed to the simultaneous inhibition of PI3K-AKT and RAS-ERK signaling pathways. purity: 98%
Catalog No. 50-257-7414