Learn More
Agilent Technologies™ RTCA CIM Plate 16
Catalog No. 5665817001
Change view
Click to view available options
Quantity:
6 Plates
6 Plates/Pk., 6 Pk./Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-658-17001 6 Plates
56-658-25001 6 Plates/Pk., 6 Pk./Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-658-17001 Supplier Agilent Technologies™ Supplier No. 5665817001
Add to Cart
Add to Cart
Observe the cell response in real-time with the RTCA CIM Plate 16.
  • Derive cell kinetics
  • Obtain continuous immune cell killing assay time points during the entire experiment
  • RTCA provides a continuous quantitative readout of proliferation rate, cell number, size/shape, and substrate attachment quality
  • PET membrane with 8 μm pore size
Specifications
Barcode Includes Barcode
Format Standard
Array 2 x 8
Color Clear, Gold
Disposable Single-use
Material PET
Volume (Metric) Well 162 ±3 μL
No. of Wells 16
Volume (Metric) Working 180 ±5 μL
Sterility Non-sterile
Surface Treatment Gold-plated
Well Shape Round, Flat Bottom (F-well)
Quantity 6 Plates