Learn More
Cole-Parmer Digi-Sense RTD Probe, 100 Ohm, 3-ft SS Braid Cable; 9" L
Supplier:  Cole-Parmer 0811797
Encompass_Preferred
RTD bendable probes are designed to measure temperature in a confined space.Maximum error for probe assembly is 1.1C + 0.12% of reading below 300F, 0.35% of reading above 300F. Alpha coefficient is 0.003850 omega/omega/C (DIN IEC 751). Resistance at ice point (0C) is 100 omega.
Catalog No. 50-196-1485