Learn More
Cole-Parmer Digi-Sense Compact RTD Probe, 100 Ohm, 3-Pin. 1.5" L, 5ft SS Braid Cable
Catalog No. 501961484 Shop All Cole-Parmer Products
Encompass_Preferred
RTD bendable probes are designed to measure temperature in a confined space.Maximum error for probe assembly is 1.1C + 0.12% of reading below 300F, 0.35% of reading above 300F. Alpha coefficient is 0.003850 omega/omega/C (DIN IEC 751). Resistance at ice point (0C) is 100 omega.
Catalog No. 50-196-1484 Supplier Cole-Parmer Supplier No. 0811796
RTD bendable probes are designed to measure temperature in a confined space.Maximum error for probe assembly is 1.1C + 0.12% of reading below 300F, 0.35% of reading above 300F. Alpha coefficient is 0.003850 omega/omega/C (DIN IEC 751). Resistance at ice point (0C) is 100 omega.