Learn More
Omega Engineering, Inc. Channel High Precision Handheld RTD with Probe Datalogger
Catalog No. NC1769453
Encompass
High accuracy RTD thermometer plus a powerful data logger. The unit can store up to 16,000 records at programmed intervals.
Catalog No. NC1769453 Supplier Omega Engineering, Inc. Supplier No. HH376
High accuracy RTD thermometer plus a powerful data logger. The unit can store up to 16,000 records at programmed intervals.