Learn More
Cole-Parmer Digi-Sense RTD Extension Cable, 3-Pin Connector Male to Female, 10ft FEP Cable
Catalog No. 501961483 Shop All Cole-Parmer Products
Encompass_Preferred
Use adapters or connectors to make your RTD probes compatible with all of our RTD thermometers. Use the extension cables for remote applications.
Catalog No. 50-196-1483 Supplier Cole-Parmer Supplier No. 0811792
Use adapters or connectors to make your RTD probes compatible with all of our RTD thermometers. Use the extension cables for remote applications.