Learn More
Cole-Parmer Digi-Sense Economical Hi-Accy RTD Probe 3 Wire 3 P" Connector, 1 / 4" Dia 20" L 1M Cable
Catalog No. 501961615 Shop All Cole-Parmer Products
Encompass_Preferred
Ideal for applications where accuracy is a factor but so is cost. Alpha coefficient is 0.003850 omega/omega/C (DIN IEC 751). Resistance at ice point (0C) is 100 omega. Time Constant: 2 seconds or greater.
Catalog No. 50-196-1615 Supplier Cole-Parmer Supplier No. 9382260
Ideal for applications where accuracy is a factor but so is cost. Alpha coefficient is 0.003850 omega/omega/C (DIN IEC 751). Resistance at ice point (0C) is 100 omega. Time Constant: 2 seconds or greater.