Learn More
Cole-Parmer Hypodermic RTD Probe, 100 Ohm, 5-ft Coil Cable; 2" L
Supplier:  Cole-Parmer 0811780
Encompass_Preferred
This probe is designed to give the same accuracy and benefits of a RTD Probe in a compact economical design. Maximum error for probe assembly is 1.1C + 0.12% of reading below 300F, 0.35% of reading above 300F. Alpha coefficient is 0.003850 //C (DIN IEC 751). Resistance at ice point (0C) is 100 .
Catalog No. 50-196-1481