Learn More
Cole-Parmer General Purpose RTD Probe, 100 Ohm, 5-ft Coil Cable; 18" L
Catalog No. NC1634315 Shop All Cole-Parmer Products
Encompass_Preferred
Maximum error for probe assembly is 1.1C + 0.12% of reading below 300F, 0.35% of reading above 300F. Alpha coefficient is 0.003850 //C (DIN IEC 751). Resistance at ice point (0C) is 100 . Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five. Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C).
Catalog No. NC1634315 Supplier Cole-Parmer Supplier No. 0811772
Maximum error for probe assembly is 1.1C + 0.12% of reading below 300F, 0.35% of reading above 300F. Alpha coefficient is 0.003850 //C (DIN IEC 751). Resistance at ice point (0C) is 100 . Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five. Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220F (104C); PVC handles are rated to 200F (93C); Stainless Steel handles are rated at 450F (232C); Mini Connectors are rated to 392F (200C).