Learn More
Cole-Parmer General Purpose RTD Probe, 100 Ohm, 5-ft Coil Cable; 10" L
Catalog No. NC1137040 Shop All Cole-Parmer Products
Encompass_Preferred
Designed to give the same accuracy and benefits of a RTD Probe in a compact economical design. Time constant : 1-3 seconds or greater. Custom designs or changes to this product are available upon request.
Catalog No. NC1137040 Supplier Cole-Parmer Supplier No. 0811770
Designed to give the same accuracy and benefits of a RTD Probe in a compact economical design. Time constant : 1-3 seconds or greater. Custom designs or changes to this product are available upon request.