Learn More
Cole-Parmer Replacement RTD Probe for Traceable Remote-Monitoring RTD Thermometer
Catalog No. NC2066673 Shop All Cole-Parmer Products
Encompass_Preferred
Extend the life of your RTF meter with these replacement RTD temperature probes. For improved performance, probes should be calibrated as a system with the meter.
Catalog No. NC2066673 Supplier Cole-Parmer Supplier No. 9876806
Extend the life of your RTF meter with these replacement RTD temperature probes. For improved performance, probes should be calibrated as a system with the meter.