Learn More
Phonetics Inc RTD Probe with 4-20mA Transmitter
SDP
Catalog No. NC1836384
Encompass_Preferred
The RTD probe with 4-20mA transmitter can be used to measure the temperature in extreme environments. The standard transmitter is calibrated to operate over the range -200° to 35°C. The sensor will need a 24VDC power supply to operate.
Catalog No. NC1836384 Supplier Phonetics Inc Supplier No. FGD0240
The RTD probe with 4-20mA transmitter can be used to measure the temperature in extreme environments. The standard transmitter is calibrated to operate over the range -200° to 35°C. The sensor will need a 24VDC power supply to operate.