Learn More
Bellco Glass, Inc. RTD Temp Sensor w/Cable For Vertical up to 6L
SDP
Catalog No. 501946275
Encompass_Preferred
RTD temperature sensor fits into a Thermowell to register thetemperature of the media inside a vessel. This sensor connects to atemperature controller with solenoid valve to activate either a heatingblanket for heating or a coil for cooling. Dimensions: 17x10x1.75" ( 43x25x4cm) 0.187x5"
Catalog No. 50-194-6275 Supplier Bellco Glass, Inc. Supplier No. 780311510
RTD temperature sensor fits into a Thermowell to register thetemperature of the media inside a vessel. This sensor connects to atemperature controller with solenoid valve to activate either a heatingblanket for heating or a coil for cooling. Dimensions: 17x10x1.75" ( 43x25x4cm) 0.187x5"