Learn More
Bellco Glass, Inc. Rtd Temp Sensor w/Cable For Vertical 8/15/36 L
SDP
Catalog No. 501946273
Encompass_Preferred
RTD temperature sensor fits into a Thermowell to register thetemperature of the media inside a vessel. This sensor connects to atemperature controller with solenoid valve to activate either a heatingblanket for heating or a coil for cooling. Dimensions: 0.187" x 10"
Catalog No. 50-194-6273 Supplier Bellco Glass, Inc. Supplier No. 780311500
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RTD temperature sensor fits into a Thermowell to register thetemperature of the media inside a vessel. This sensor connects to atemperature controller with solenoid valve to activate either a heatingblanket for heating or a coil for cooling. Dimensions: 0.187" x 10"