Learn More
Bellco Glass, Inc. Rtd Temp Sensor w/Cable For Angled Sidearms
SDP
Catalog No. 501946274
Encompass_Preferred
RTD temperature sensor fits into a Thermowell to register thetemperature of the media inside a vessel. This sensor connects to atemperature controller with solenoid valve to activate either a heatingblanket for heating or a coil for cooling. Dimensions: 17x10x1.75" ( 43x25x4cm ) 0.87 x 3/4"
Catalog No. 50-194-6274 Supplier Bellco Glass, Inc. Supplier No. 780311505
RTD temperature sensor fits into a Thermowell to register thetemperature of the media inside a vessel. This sensor connects to atemperature controller with solenoid valve to activate either a heatingblanket for heating or a coil for cooling. Dimensions: 17x10x1.75" ( 43x25x4cm ) 0.87 x 3/4"