Learn More
Bellco Glass, Inc. RTD Temperature Sensor,3/4 BCC3 Controller & Angled Side Arms
SDP
Catalog No. 501946518
Encompass_Preferred
Rtd temperature sensor fits into a Thermowell to register the temperature of the media inside a vessel. This sensor connects to a temperature controller with solenoid valve to activate either a heating blanket for heating or a coil for cooling. MADE IN THE USA
Catalog No. 50-194-6518 Supplier Bellco Glass, Inc. Supplier No. 791900006
Rtd temperature sensor fits into a Thermowell to register the temperature of the media inside a vessel. This sensor connects to a temperature controller with solenoid valve to activate either a heating blanket for heating or a coil for cooling. MADE IN THE USA