Learn More
Cole-Parmer Digi-Sense RTD Thermometer, NIST-Traceable Calibration
Catalog No. NC1133934 Shop All Cole-Parmer Products
Encompass_Preferred
The Digi-Sense Professional Single-Input Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours. Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you hundreds of dollars and extra waiting time to calibrate separately.
Catalog No. NC1133934 Supplier Cole-Parmer Supplier No. 2025095
The Digi-Sense Professional Single-Input Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours. Digi-Sense instrument are fully tested and calibrated to NIST-traceable standards, saving you hundreds of dollars and extra waiting time to calibrate separately.