Learn More
Cayman Chemical RTDLdLRTYTLtrIfluoroACTAt 10mg
SDP
Catalog No. 502915828
Encompass_Preferred
An αVβ6 integrin ligand; selectively inhibits the binding of fibronectin to αVβ6 integrin over vitronectin to αVβ3 or αVβ5 and fibrinogen to αIIβ3 (IC50s = 0.02, >50, >50, and >50 µM, respectively); inhibits the binding of HT-29 colon cancer cells to fibronectin
Catalog No. 50-291-5828 Supplier Cayman Chemical Supplier No. 3745510 MG
An αVβ6 integrin ligand; selectively inhibits the binding of fibronectin to αVβ6 integrin over vitronectin to αVβ3 or αVβ5 and fibrinogen to αIIβ3 (IC50s = 0.02, >50, >50, and >50 µM, respectively); inhibits the binding of HT-29 colon cancer cells to fibronectin