Learn More
Lonza Walkersville RtEBM Retinal Epithelial Cell Basal Medium
Catalog No. NC1583479
Encompass
RtEBM is a Retinal Epithelial Cell basal medium sold separately. This can be used with 1 kit of 195407 (retinal epithelial cell growth medium SingleQuots Supplements and Growth Factors). The full RtEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 195409.RtEGM Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum. Applications:Age-related macular degenerationRetinitis pigmentosaDiabetic retinopathyPhotoreceptor degenerationGene expressionCellular interactionsContent:1 x Bottle Basal Medium 200 mL
Catalog No. NC1583479 Supplier Lonza Walkersville Supplier No. 00195406
RtEBM is a Retinal Epithelial Cell basal medium sold separately. This can be used with 1 kit of 195407 (retinal epithelial cell growth medium SingleQuots Supplements and Growth Factors). The full RtEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 195409.RtEGM Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum. Applications:Age-related macular degenerationRetinitis pigmentosaDiabetic retinopathyPhotoreceptor degenerationGene expressionCellular interactionsContent:1 x Bottle Basal Medium 200 mL