Learn More
Lonza Walkersville RtEGM Retinal Pigment Epithelial Cell Growth Medium BulletKit
Catalog No. NC0718053
Encompass
RtEGM Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum.Applications:Age-related macular degenerationRetinitis pigmentosaDiabetic retinopathyPhotoreceptor degenerationGene expressionCellular interactions Content:1 x RtEBM Basal Medium (00195406), 200 mL1 x RtEGM SingleQuots Supplement Pack (00195407) containing:1 x Bottle L-Glutamine, 4.00 mL1 x Gray Cap Vial with hFGF-B, 1.00 mL1 x Bottle FBS, 4.00 mL1 x Red Cap Vial with GA-1000, 0.20 mL
Catalog No. NC0718053 Supplier Lonza Walkersville Supplier No. 00195409
RtEGM Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum.Applications:Age-related macular degenerationRetinitis pigmentosaDiabetic retinopathyPhotoreceptor degenerationGene expressionCellular interactions Content:1 x RtEBM Basal Medium (00195406), 200 mL1 x RtEGM SingleQuots Supplement Pack (00195407) containing:1 x Bottle L-Glutamine, 4.00 mL1 x Gray Cap Vial with hFGF-B, 1.00 mL1 x Bottle FBS, 4.00 mL1 x Red Cap Vial with GA-1000, 0.20 mL