Learn More
Lonza Walkersville RtEGM Retinal Epithelial Cell Growth Medium SingleQuots Supplements and Growth Factors
Catalog No. NC1583487
Encompass
RtGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the RtEGM Retinal Epithelial Cell Growth Medium BulletKit (Catalog Number 195409). The full RtEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 195409.RtEGM Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum.Applications:Age-related macular degenerationRetinitis pigmentosaDiabetic retinopathyPhotoreceptor degenerationGene expressionCellular interactionsContent:1 x Bottle with L-Glutamine 4.00 mL, 1 x Gray Cap Vial with hFGF-B 1.00 mL, 1 x Bottle FBS 4.00 mL, 1 x Red Cap Vial with GA-1000 0.20 mL
Catalog No. NC1583487 Supplier Lonza Walkersville Supplier No. 00195407
RtGM SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the RtEGM Retinal Epithelial Cell Growth Medium BulletKit (Catalog Number 195409). The full RtEGM BulletKit consisting of basal medium and supplements can be purchased together using the catalog number 195409.RtEGM Retinal Pigment Epithelial Cell Growth Medium is optimized and performance tested for growing Human RPE cells. It contains 2% serum.Applications:Age-related macular degenerationRetinitis pigmentosaDiabetic retinopathyPhotoreceptor degenerationGene expressionCellular interactionsContent:1 x Bottle with L-Glutamine 4.00 mL, 1 x Gray Cap Vial with hFGF-B 1.00 mL, 1 x Bottle FBS 4.00 mL, 1 x Red Cap Vial with GA-1000 0.20 mL