Learn More
Medchemexpress LLC RTI-13951-33 hydrochloride | 98.06% | 532.50 | 1 ML
SDP
Catalog No. 5000437590
Encompass_Preferred
RTI-13951-33 hydrochloride is a potent, selective, and brain-penetrant GPR88 agonist. It has an EC50 of 25 nM in GPR88 cAMP functional assay and reduces alcohol reinforcement and intake behaviors in rats. This compound elevates [35S]-GTPγS binding in mouse striatal membranes and demonstrates sufficient brain penetration.
  • Potent and selective GPR88 agonist
  • Brain-penetrant properties
  • Reduces alcohol reinforcement and intake behaviors
  • Elevates GTPγS binding in mouse striatal membranes
Catalog No. 50-004-37590 Supplier Medchemexpress LLC Supplier No. HY112612A10MM/1ML
RTI-13951-33 hydrochloride is a potent, selective, and brain-penetrant GPR88 agonist. It has an EC50 of 25 nM in GPR88 cAMP functional assay and reduces alcohol reinforcement and intake behaviors in rats. This compound elevates [35S]-GTPγS binding in mouse striatal membranes and demonstrates sufficient brain penetration.
  • Potent and selective GPR88 agonist
  • Brain-penetrant properties
  • Reduces alcohol reinforcement and intake behaviors
  • Elevates GTPγS binding in mouse striatal membranes