Learn More
Medchemexpress LLC RTI-7470-44 | 825658-63-7 | 99.9% | 449.84 | 5 MG
SDP
Catalog No. 5000362336
Encompass_Preferred
RTI-7470-44 is a potent, selective, and blood-brain barrier (BBB) penetrant human trace amine-associated receptor subtype 1 (hTAAR1) antagonist. It exhibits moderate metabolic stability and a favorable preliminary off-target profile.
  • Potent and selective hTAAR1 antagonist
  • Blood-brain barrier penetrant
  • Increases spontaneous firing rate of mouse VTA dopaminergic neurons
  • Used for researching schizophrenia, agent addiction, and Parkinson's disease
Catalog No. 50-003-62336 Supplier Medchemexpress LLC Supplier No. HY1473195MG
RTI-7470-44 is a potent, selective, and blood-brain barrier (BBB) penetrant human trace amine-associated receptor subtype 1 (hTAAR1) antagonist. It exhibits moderate metabolic stability and a favorable preliminary off-target profile.
  • Potent and selective hTAAR1 antagonist
  • Blood-brain barrier penetrant
  • Increases spontaneous firing rate of mouse VTA dopaminergic neurons
  • Used for researching schizophrenia, agent addiction, and Parkinson's disease