Learn More
Medchemexpress LLC RTI-7470-44 | 825658-63-7 | 99.9% | C19H11ClF3N5OS | 10MG
SDP
Catalog No. 5000259543
Encompass_Preferred
RTI-7470-44 is a potent, selective, blood-brain barrier-penetrant antagonist of human trace amine-associated receptor 1 (hTAAR1) intended for preclinical research. It shows low-nanomolar potency and is used in electrophysiology and in vivo studies to probe dopaminergic neuron activity relevant to schizophrenia, substance addiction, and Parkinson's disease.
  • Potent, selective hTAAR1 antagonist.
  • Blood-brain barrier penetrant, enabling CNS studies.
  • Low-nanomolar potency against hTAAR1.
  • High purity suitable for biochemical and in vivo experiments.
  • Applicable to electrophysiology and behavioral models.
Catalog No. 50-002-59543 Supplier Medchemexpress LLC Supplier No. HY14731910MG
RTI-7470-44 is a potent, selective, blood-brain barrier-penetrant antagonist of human trace amine-associated receptor 1 (hTAAR1) intended for preclinical research. It shows low-nanomolar potency and is used in electrophysiology and in vivo studies to probe dopaminergic neuron activity relevant to schizophrenia, substance addiction, and Parkinson's disease.
  • Potent, selective hTAAR1 antagonist.
  • Blood-brain barrier penetrant, enabling CNS studies.
  • Low-nanomolar potency against hTAAR1.
  • High purity suitable for biochemical and in vivo experiments.
  • Applicable to electrophysiology and behavioral models.