Learn More
Aviva Systems Biology Corp RTN1 Antibody - middle region (ARP49623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RTN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RTN1. Alternate names: MGC133250, NSPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-60126