Learn More
Aviva Systems Biology Corp RTN4IP1 Antibody - middle region (ARP60655_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60655_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RTN4IP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RTN4IP1. Alternate names: MGC12934, NIMPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-116