Learn More
Aviva Systems Biology Corp RTP1 Antibody - N-terminal region (ARP65494_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65494_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RTP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human RTP1. Alternate names: MGC35450ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-508