Learn More
Aviva Systems Biology Corp RTP4 Antibody - middle region (ARP57612_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57612_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RTP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RTP4. Alternate names: IFRG28ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62871