Learn More
Chemglass Life Sciences Bump Trap, 100mL, Rotary Evaporator, Modified, 24/40 Top Outer, 14/20 Lower Inner
SDP
Catalog No. CG132101
Encompass_Preferred
Bump Trap, 100mL, Rotary Evaporator, Modified, 24/40 Top Outer, 14/20 Lower Inner. Similar to CG-1320, but with the addition of two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Traps have a 24/40 top outer joint and the listed size of lower inner joint.
Catalog No. CG132101 Supplier Chemglass Life Sciences Supplier No. CG132101
Bump Trap, 100mL, Rotary Evaporator, Modified, 24/40 Top Outer, 14/20 Lower Inner. Similar to CG-1320, but with the addition of two 4mm drain holes at the base of the inner tube to allow solvent to drain back into the evaporation flask. Traps have a 24/40 top outer joint and the listed size of lower inner joint.