Learn More
Chemglass Life Sciences Bump Trap, 500mL, Rotary Evaporator, Modified, 24/40 Top Outer, 24/40 Lower Inner
SDP
Supplier:  Chemglass Life Sciences CG132209
Encompass_Preferred
Bump Trap, 500mL, Rotary Evaporator, Modified, 24/40 Top Outer, 24/40 Lower Inner. Trap provides expanded volume for reactions that have a tendency to foam excessively. The inner vapor tube is sealed to the top of the trap to prevent entry into the condenser. Traps have a 24/40 top outer joint and the listed size of lower inner joint.
Catalog No. CG132209