Learn More
MSC Fiberlock Disinfectant/Sanitizer, Ready-to-Use, EPA Registered, For Porous/Non-Porous Surfaces
Catalog No. 502839922
Encompass
This ready-to-use formulation is designed for use on various surfaces subject to microbial contamination, making it suitable for flooding and mold remediation. It is particularly useful for treating wood framing, carpets, and other porous and nonporous surfaces as part of a complete mold remediation project.
  • EPA registered on porous and non-porous surfaces.
  • Registered for sewer/black water remediation.
  • Kills over 120 organisms.
  • Hospital grade disinfectant.
  • Ready-to-use formula.
  • Features antimicrobial, deodorizing, disinfecting, and sanitizing properties.
Catalog No. 50-283-9922 Supplier MSC Supplier No. 90259375
This ready-to-use formulation is designed for use on various surfaces subject to microbial contamination, making it suitable for flooding and mold remediation. It is particularly useful for treating wood framing, carpets, and other porous and nonporous surfaces as part of a complete mold remediation project.
  • EPA registered on porous and non-porous surfaces.
  • Registered for sewer/black water remediation.
  • Kills over 120 organisms.
  • Hospital grade disinfectant.
  • Ready-to-use formula.
  • Features antimicrobial, deodorizing, disinfecting, and sanitizing properties.