Learn More
MSC Fiberlock Disinfectant/Sanitizer, Ready-to-Use, 5 Gallon, EPA Registered
Catalog No. 502839899
Encompass
Shockwave RTU is a convenient, ready-to-use disinfectant and sanitizer, ideal for situations without a clean water source and for flood and mold remediation. This all-in-one liquid is designed for use on various porous and non-porous surfaces, effectively treating wood framing, carpets, and other materials. It boasts over 120 organism kill claims and features a fresh laundry scent.
  • EPA registered for porous and non-porous surfaces
  • Registered for sewer/black water remediation
  • Effective against over 120 organism kill claims
  • Ready-to-use formulation
  • Features a fresh laundry scent
  • Suitable for flood and mold remediation
  • Can be used on wood framing, carpets, and other surfaces
Catalog No. 50-283-9899 Supplier MSC Supplier No. 90259383
Shockwave RTU is a convenient, ready-to-use disinfectant and sanitizer, ideal for situations without a clean water source and for flood and mold remediation. This all-in-one liquid is designed for use on various porous and non-porous surfaces, effectively treating wood framing, carpets, and other materials. It boasts over 120 organism kill claims and features a fresh laundry scent.
  • EPA registered for porous and non-porous surfaces
  • Registered for sewer/black water remediation
  • Effective against over 120 organism kill claims
  • Ready-to-use formulation
  • Features a fresh laundry scent
  • Suitable for flood and mold remediation
  • Can be used on wood framing, carpets, and other surfaces