Learn More
Bellco Glass, Inc. RTV Red Glue #106 2.8 OZ
SDP
Catalog No. 501944599
Encompass_Preferred
Rtv red glue #106 208ozUsed for replacing silicone pads in pipette cans. Easy to use nozzle and adheres to stainless steel aluminum, glass, and rigid plastics. Curing time: 24 hours Temperature range: 60-260 degree Celsius
Catalog No. 50-194-4599 Supplier Bellco Glass, Inc. Supplier No. 121700028
Rtv red glue #106 208ozUsed for replacing silicone pads in pipette cans. Easy to use nozzle and adheres to stainless steel aluminum, glass, and rigid plastics. Curing time: 24 hours Temperature range: 60-260 degree Celsius