Learn More
Accuflow Systems 30x60" Mobile RTW Lab Table with phenolic resin top
Catalog No. 501489317
Encompass_Preferred
30x60" Mobile with casters RTW Table by Production Basics. Almond color 13-14 Gauge Steel Frame with black phenolic resin top. Height Adjustment 30-36", Weight Capactiy 750lbs. Ships knocked-down within 10-15 business days.
Catalog No. 50-148-9317 Supplier Accuflow Systems Supplier No. 3738
30x60" Mobile with casters RTW Table by Production Basics. Almond color 13-14 Gauge Steel Frame with black phenolic resin top. Height Adjustment 30-36", Weight Capactiy 750lbs. Ships knocked-down within 10-15 business days.