Learn More
Medchemexpress LLC RU 24969 succinate | 107008-28-6 | 99.6% | 346.38 | 5 MG
SDP
Catalog No. 5000392339
Encompass_Preferred
RU 24969 succinate is a 5-HT receptor agonist with Ki values of 0.38 and 2.5 nM for 5-HT1B and 5-HT1A, respectively. It decreases fluid consumption and increases forward locomotion. It can be used for the research of neurological diseases.
  • 5-HT receptor agonist with Ki values of 0.38 and 2.5 nM for 5-HT1B and 5-HT1A, respectively.
  • Decreases fluid consumption.
  • Increases forward locomotion.
  • Can be used for the research of neurological disease.
  • Potentiates addictive agent-induced effects.
  • Dose-dependently decreases water consumption in water deprived rats.
  • Increased cocaine-induced elevation of nucleus accumbens (NAcc) dopamine DA levels and reduction of ventral tegmental area (VTA) GABA levels in male Wistar rats.
Catalog No. 50-003-92339 Supplier Medchemexpress LLC Supplier No. HY16688A5MG
RU 24969 succinate is a 5-HT receptor agonist with Ki values of 0.38 and 2.5 nM for 5-HT1B and 5-HT1A, respectively. It decreases fluid consumption and increases forward locomotion. It can be used for the research of neurological diseases.
  • 5-HT receptor agonist with Ki values of 0.38 and 2.5 nM for 5-HT1B and 5-HT1A, respectively.
  • Decreases fluid consumption.
  • Increases forward locomotion.
  • Can be used for the research of neurological disease.
  • Potentiates addictive agent-induced effects.
  • Dose-dependently decreases water consumption in water deprived rats.
  • Increased cocaine-induced elevation of nucleus accumbens (NAcc) dopamine DA levels and reduction of ventral tegmental area (VTA) GABA levels in male Wistar rats.