Learn More
Medchemexpress LLC RU 24969 succinate | 107008-28-6 | 99.6% | 346.38 | 50 MG
SDP
Catalog No. 5000391845
Encompass_Preferred
RU 24969 succinate is a 5-HT receptor agonist with Ki values of 0.38 and 2.5 nM for 5-HT1B and 5-HT1A, respectively. This compound decreases fluid consumption and increases forward locomotion, making it suitable for research into neurological diseases.
  • Acts as a 5-HT receptor agonist with Ki values of 0.38 nM for 5-HT1B and 2.5 nM for 5-HT1A.
  • Decreases fluid consumption and increases forward locomotion.
  • Can be used for the research of neurological disease.
  • Dose-dependently inhibits K+ evoked efflux of tritium (30 nM-1 μM) with a pD2 value of 7.45 for the maximum effect at 1 μM.
  • Potentiates addictive agent-induced effects (1 and 3 mg/kg; s.c., once) and dose-dependently decreases water consumption in water deprived rats (0.03-3.0 mg/kg; s.c., once).
Catalog No. 50-003-91845 Supplier Medchemexpress LLC Supplier No. HY16688A50MG
RU 24969 succinate is a 5-HT receptor agonist with Ki values of 0.38 and 2.5 nM for 5-HT1B and 5-HT1A, respectively. This compound decreases fluid consumption and increases forward locomotion, making it suitable for research into neurological diseases.
  • Acts as a 5-HT receptor agonist with Ki values of 0.38 nM for 5-HT1B and 2.5 nM for 5-HT1A.
  • Decreases fluid consumption and increases forward locomotion.
  • Can be used for the research of neurological disease.
  • Dose-dependently inhibits K+ evoked efflux of tritium (30 nM-1 μM) with a pD2 value of 7.45 for the maximum effect at 1 μM.
  • Potentiates addictive agent-induced effects (1 and 3 mg/kg; s.c., once) and dose-dependently decreases water consumption in water deprived rats (0.03-3.0 mg/kg; s.c., once).