Learn More
Medchemexpress LLC Ru 24969 succinate | 107008-28-6 | 99.6% | 346.38 | C18H22N2O5 | 10 MG
SDP
Catalog No. 5000289251
Encompass_Preferred
RU 24969 succinate is the succinate salt of RU 24969, a research-grade 5-HT receptor agonist with preferential activity at 5-HT1B and appreciable affinity for 5-HT1A. It is provided as a high-purity chemical for pharmacology and behavioral studies, with supporting documentation for research use.
  • High purity (99.58%)
  • Succinate salt form
  • Suitable for in vitro and in vivo 5-HT receptor research
  • Available in small research sizes, including 10 mg
  • Storage: powder -20°C (3 years) or 4°C (2 years)
  • Datasheet, COA, and SDS available
Catalog No. 50-002-89251 Supplier Medchemexpress LLC Supplier No. HY16688A10MG
RU 24969 succinate is the succinate salt of RU 24969, a research-grade 5-HT receptor agonist with preferential activity at 5-HT1B and appreciable affinity for 5-HT1A. It is provided as a high-purity chemical for pharmacology and behavioral studies, with supporting documentation for research use.
  • High purity (99.58%)
  • Succinate salt form
  • Suitable for in vitro and in vivo 5-HT receptor research
  • Available in small research sizes, including 10 mg
  • Storage: powder -20°C (3 years) or 4°C (2 years)
  • Datasheet, COA, and SDS available