Learn More
Medchemexpress LLC HY-119039 5mg Medchemexpress, RU-301 CAS:1110873-99-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1190395MG
Encompass_Preferred
Medchemexpress, HY-119039 5mg RU-301 CAS:1110873-99-8 RU-301 is a pan-TAM receptor inhibitor, exerts pan-TAM inhibitory activity by binding at the interface between Gas6 and the Ig1 domain of the respective TAMs with Kd and IC50 values of 12 μM and 10 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9513