Learn More
Medchemexpress LLC Ru-302 | 1182129-77-6 | HY-124066 | 98.7% | 475.53 | 5 MG
SDP
Catalog No. 5000364286
Encompass_Preferred
RU-302 is a pan TAM inhibitor that blocks the interface between the TAM Ig1 ectodomain and the Gas6 Lg domain. It effectively blocks Gas6-inducible Axl receptor activation with a low micromolar IC50 in cell assays and suppresses lung cancer tumor growth.
  • Pan TAM inhibitor
  • Blocks the interface between the TAM Ig1 ectodomain and the Gas6 Lg domain
  • Effectively blocks Gas6-inducible Axl receptor activation with a low micromolar IC50 in cell assays
  • Suppresses lung cancer tumor growth
  • Targets Axl receptor
  • Relevant to cancer, lung cancer, and protein tyrosine kinase/RTK research
Catalog No. 50-003-64286 Supplier Medchemexpress LLC Supplier No. 5MGHY124066
RU-302 is a pan TAM inhibitor that blocks the interface between the TAM Ig1 ectodomain and the Gas6 Lg domain. It effectively blocks Gas6-inducible Axl receptor activation with a low micromolar IC50 in cell assays and suppresses lung cancer tumor growth.
  • Pan TAM inhibitor
  • Blocks the interface between the TAM Ig1 ectodomain and the Gas6 Lg domain
  • Effectively blocks Gas6-inducible Axl receptor activation with a low micromolar IC50 in cell assays
  • Suppresses lung cancer tumor growth
  • Targets Axl receptor
  • Relevant to cancer, lung cancer, and protein tyrosine kinase/RTK research