Learn More
Medchemexpress LLC RU-302 (pan-TAM inhibitor) | 1182129-77-6 | Not found | 98.7% | 475.53 g/mol | C24H24F3N3O2S | 50MG
SDP
Catalog No. 5000235312
Encompass_Preferred
RU-302 is a small-molecule pan-TAM (Tyro3, Axl, Mer) inhibitor used in laboratory research to block the interface between the TAM Ig1 ectodomain and the Gas6 LG domain. It is provided as a powder or DMSO solution for in vitro studies and is intended for research use only; not for human or diagnostic use. CAS: 1182129-77-6.
Catalog No. 50-002-35312 Supplier Medchemexpress LLC Supplier No. HY12406650MG
RU-302 is a small-molecule pan-TAM (Tyro3, Axl, Mer) inhibitor used in laboratory research to block the interface between the TAM Ig1 ectodomain and the Gas6 LG domain. It is provided as a powder or DMSO solution for in vitro studies and is intended for research use only; not for human or diagnostic use. CAS: 1182129-77-6.