Learn More
Medchemexpress LLC RU.521 100mg | 2262452-06-0 | 100 MG
SDP
Catalog No. 5000229871
Encompass_Preferred
RU.521 is a small-molecule inhibitor of cyclic GMP-AMP synthase (cGAS) used in research to suppress cGAS-mediated interferon signaling. It is supplied as a high-purity solid and is provided in multiple sizes and formulations for in vitro and cellular studies.
  • High purity (99.95%) for reproducible results.
  • Potent, selective inhibition of cGAS in cellular assays.
  • Available in multiple sizes and DMSO solution formats for flexibility.
  • Stable as a powder at -20°C for long-term storage.
  • Molecular weight 415.23; formula C19H12Cl2N4O3.
Catalog No. 50-002-29871 Supplier Medchemexpress LLC Supplier No. HY114180100MG
RU.521 is a small-molecule inhibitor of cyclic GMP-AMP synthase (cGAS) used in research to suppress cGAS-mediated interferon signaling. It is supplied as a high-purity solid and is provided in multiple sizes and formulations for in vitro and cellular studies.
  • High purity (99.95%) for reproducible results.
  • Potent, selective inhibition of cGAS in cellular assays.
  • Available in multiple sizes and DMSO solution formats for flexibility.
  • Stable as a powder at -20°C for long-term storage.
  • Molecular weight 415.23; formula C19H12Cl2N4O3.