Learn More
TARGETMOL CHEMICALS INC RU.521 10MG
SDP
Catalog No. 502579754
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RU.521 (RU3205217) is a potent dsDNA inhibitor (IC50 700 nM) inhibition of the dsDNA pathway by RU.521 requires the presence of cGAS. purity: 99%
Catalog No. 50-257-9754 Supplier TARGETMOL CHEMICALS INC Supplier No. T548610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RU.521 (RU3205217) is a potent dsDNA inhibitor (IC50 700 nM) inhibition of the dsDNA pathway by RU.521 requires the presence of cGAS. purity: 99%