Learn More
Medchemexpress LLC RU.521 10MG
SDP
Catalog No. NC1743650
Encompass_Preferred
RU.521 (RU320521) is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor and inhibits cGAS-mediated interferon upregulation | CAS 2262452-06-0 | SMILES O=C1OC(C2=C(O)N(C3=NC4=CC=C(Cl)C(Cl)=C4N3)N=C2C)C5=C1C=CC=C5
Catalog No. NC1743650 Supplier Medchemexpress LLC Supplier No. HY11418010MG
RU.521 (RU320521) is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor and inhibits cGAS-mediated interferon upregulation | CAS 2262452-06-0 | SMILES O=C1OC(C2=C(O)N(C3=NC4=CC=C(Cl)C(Cl)=C4N3)N=C2C)C5=C1C=CC=C5