Learn More
Medchemexpress LLC RU-521 | 2262452-06-0 | 100.0% | 415.23 | 50 MG
SDP
Catalog No. 5000365947
Encompass_Preferred
RU.521 is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor. It inhibits cGAS-mediated interferon upregulation and suppresses dsDNA-activated reporter activity.
  • Inhibits cGAS-mediated interferon upregulation.
  • Suppresses dsDNA-activated reporter activity with an IC50 of 0.7 μM.
  • Reduces constitutive expression of interferon in macrophages from a mouse model of Aicardi-Goutières syndrome (AGS).
  • Reduces symptoms from sepsis in mice, increasing cardiac function and decreasing inflammatory responses.
Catalog No. 50-003-65947 Supplier Medchemexpress LLC Supplier No. 50MGHY114180
RU.521 is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor. It inhibits cGAS-mediated interferon upregulation and suppresses dsDNA-activated reporter activity.
  • Inhibits cGAS-mediated interferon upregulation.
  • Suppresses dsDNA-activated reporter activity with an IC50 of 0.7 μM.
  • Reduces constitutive expression of interferon in macrophages from a mouse model of Aicardi-Goutières syndrome (AGS).
  • Reduces symptoms from sepsis in mice, increasing cardiac function and decreasing inflammatory responses.