Learn More
Medchemexpress LLC RU.521 | 25MG
SDP
Catalog No. 5000226686
Encompass_Preferred
RU 521 (RU320521) is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor and inhibits cGAS-mediated interferon upregulation RU 521 suppresses dsDNA-activated reporter activity with an IC50 of 0 7 M RU 521 reduces constitutive expression of interferon in macrophages from a mouse model of Aicardi-Gouti res syndrome (AGS)[1]
Catalog No. 50-002-26686 Supplier Medchemexpress LLC Supplier No. HY11418025MG
RU 521 (RU320521) is a potent and selective cyclic GMP-AMP synthase (cGAS) inhibitor and inhibits cGAS-mediated interferon upregulation RU 521 suppresses dsDNA-activated reporter activity with an IC50 of 0 7 M RU 521 reduces constitutive expression of interferon in macrophages from a mouse model of Aicardi-Gouti res syndrome (AGS)[1]