Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC RU.521 500mg | 2262452-06-0 | 500 MG
SDP

Catalog No. 5000226684
Encompass_Preferred

RU.521 is a potent, selective small-molecule inhibitor of cyclic GMP-AMP synthase (cGAS) provided as a solid research chemical for in vitro and in vivo pathway and interferon-response studies.

  • Potent and selective cGAS inhibitor.
  • High purity suitable for research use.
  • Chemical formula C19H12Cl2N4O3; molecular weight 415.23.
  • Soluble in DMSO; in vivo formulation examples are reported.
  • Available in multiple package sizes, including a 500 mg pack.
  • Store powder at -20°C for long-term stability; in solvent storage recommendations provided.

Catalog No. 50-002-26684 Supplier Medchemexpress LLC Supplier No. HY114180500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RU.521 is a potent, selective small-molecule inhibitor of cyclic GMP-AMP synthase (cGAS) provided as a solid research chemical for in vitro and in vivo pathway and interferon-response studies.

  • Potent and selective cGAS inhibitor.
  • High purity suitable for research use.
  • Chemical formula C19H12Cl2N4O3; molecular weight 415.23.
  • Soluble in DMSO; in vivo formulation examples are reported.
  • Available in multiple package sizes, including a 500 mg pack.
  • Store powder at -20°C for long-term stability; in solvent storage recommendations provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.