Learn More
Selleck Chemical LLC RU.521 5mg
SDP
Supplier:  Selleck Chemical LLC S68415MG
Encompass_Preferred
RU.521 (RU320521) is an active, potent and selective inhibitor of cyclic GMP-AMP synthase (cGAS) that inhibits the cGAS-mediated signaling with IC50 of 700 nM. RU.521 binds to the cGAS/dsDNA complex with Kd of 36.2 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3227