Learn More
Medchemexpress LLC Ru(bpy)2(mcbpy-O-Su-ester)(PF6)2 | 136724-73-7 | MFCD02683415 | 97.0% | 1,014.66 g/mol | C36H29F12N7O4P2Ru | 1 MG
SDP
Catalog No. 5000283638
Encompass_Preferred
Ru(bpy)2(mcbpy-O-Su-ester)(PF6)2 is a ruthenium-based activated ester dye used for covalent labeling of amine-containing biomolecules and fluorescence assays. It is supplied as the hexafluorophosphate salt, a brown-red solid (MW 1,014.66 g/mol) with reported purity of 97%.
  • Activated ester for acylation of primary amines.
  • Long fluorescence lifetime and high photostability.
  • Suitable for labeling proteins and peptides for fluorescence detection.
  • Brown-red solid; store at -20°C, protected from moisture and light.
  • High reported purity for reliable labeling performance.
Catalog No. 50-002-83638 Supplier Medchemexpress LLC Supplier No. HYW1277161MG
Ru(bpy)2(mcbpy-O-Su-ester)(PF6)2 is a ruthenium-based activated ester dye used for covalent labeling of amine-containing biomolecules and fluorescence assays. It is supplied as the hexafluorophosphate salt, a brown-red solid (MW 1,014.66 g/mol) with reported purity of 97%.
  • Activated ester for acylation of primary amines.
  • Long fluorescence lifetime and high photostability.
  • Suitable for labeling proteins and peptides for fluorescence detection.
  • Brown-red solid; store at -20°C, protected from moisture and light.
  • High reported purity for reliable labeling performance.