Learn More
Strem, An Ascensus Company [Ru(Me5Cp)(MeCN)3]PF6 50mg
SDP
Supplier:  Strem, An Ascensus Company 44788050MG
Encompass
CAS# 99604-67-8. 50mg. Tris(acetonitrile)pentamethylcyclopentadienylruthenium(II) hexafluorophosphate, min. 98%. MFCD04117715. Molecular Weight: 504.42. Molecular Formula: [Ru(C10H15)(CH3CN)3]+PF6ae. Color/form: yellow orange pwdr. Strem# 44-7880. http://www.strem.com/catalog/v/44-7880/
Catalog No. 50-203-4129